Product Details

SNP ID
rs104894533
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:8801827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTACCAAAGAAATGGATGACTTCC[G/T]ACAAAAATTGAGGCAGAAGATCAAA
Phenotype
MIM: 601785
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PMM2 PubMed Links

Gene Details

Gene
PMM2
Gene Name
phosphomannomutase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000303.2 Intron NP_000294.1
XM_005255374.4 Intron XP_005255431.1
Gene
TMEM186
Gene Name
transmembrane protein 186
There are no transcripts associated with this gene.

View Full Product Details