Product Details

SNP ID
rs17625219
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:152843013 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCCAGGGAGCTGAAAAGCCAGG[C/T]AGGAGATGCTTGGGGTAGAAAGAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LCE6A PubMed Links

Gene Details

Gene
LCE6A
Gene Name
late cornified envelope 6A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128600.1 Intron NP_001122072.1
XM_017001327.1 Intron XP_016856816.1

View Full Product Details