Product Details

SNP ID
rs17711242
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:85147038 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTACACTTGGTTCCTTAAATATCA[G/T]AGGTGAGATGTTAACAGTTCTAGGT
Phenotype
MIM: 604652
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TCF7L1 PubMed Links

Gene Details

Gene
TCF7L1
Gene Name
transcription factor 7 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031283.2 Intron NP_112573.1
XM_006712109.2 Intron XP_006712172.1

View Full Product Details