Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001144881.1 | 2999 | UTR 3 | NP_001138353.1 | ||
NM_001144882.1 | 2999 | UTR 3 | NP_001138354.1 | ||
NM_001144883.1 | 2999 | UTR 3 | NP_001138355.1 | ||
NM_153026.2 | 2999 | UTR 3 | NP_694571.2 | ||
XM_011537946.1 | 2999 | UTR 3 | XP_011536248.1 | ||
XM_011537947.2 | 2999 | UTR 3 | XP_011536249.1 | ||
XM_017018838.1 | 2999 | UTR 3 | XP_016874327.1 | ||
XM_017018839.1 | 2999 | UTR 3 | XP_016874328.1 | ||
XM_017018840.1 | 2999 | UTR 3 | XP_016874329.1 |