Product Details

SNP ID
rs17726541
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150494288 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCATATGTAGTCTGAGGGGGCTGA[A/G]TGGCCTTAACTGCTATCAATCCAGT
Phenotype
MIM: 600853
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC102546298 PubMed Links

Gene Details

Gene
LOC102546298
Gene Name
uncharacterized LOC102546298
There are no transcripts associated with this gene.

Gene
NDST1
Gene Name
N-deacetylase and N-sulfotransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301063.1 Intron NP_001287992.1
NM_001543.4 Intron NP_001534.1
XM_005268433.1 Intron XP_005268490.1
XM_005268434.2 Intron XP_005268491.1
XM_005268435.4 Intron XP_005268492.1
XM_005268436.3 Intron XP_005268493.1
XM_005268437.4 Intron XP_005268494.1
XM_005268438.1 Intron XP_005268495.1
XM_005268439.1 Intron XP_005268496.1
XM_005268441.1 Intron XP_005268498.1
XM_005268442.3 Intron XP_005268499.1
XM_006714782.1 Intron XP_006714845.1
XM_006714783.1 Intron XP_006714846.1
XM_011537638.2 Intron XP_011535940.1
XM_017009427.1 Intron XP_016864916.1
XM_017009428.1 Intron XP_016864917.1
XM_017009429.1 Intron XP_016864918.1
XM_017009430.1 Intron XP_016864919.1
XM_017009431.1 Intron XP_016864920.1
XM_017009432.1 Intron XP_016864921.1
XM_017009433.1 Intron XP_016864922.1

View Full Product Details