Product Details

SNP ID
rs17106462
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:51245843 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAAGGAAGGTAATACATAAAAGAC[A/G]TATTTGCAAACGACTTGAGGCAAGA
Phenotype
MIM: 612598
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF11 PubMed Links

Gene Details

Gene
RNF11
Gene Name
ring finger protein 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014372.4 Intron NP_055187.1

View Full Product Details