Product Details

SNP ID
rs16868618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:16455250 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGAATCTTTTGTGTAAAAAGAC[C/T]ATGAACCAGATACTATAAATATTGT
Phenotype
MIM: 608694
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF622 PubMed Links

Gene Details

Gene
ZNF622
Gene Name
zinc finger protein 622
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033414.2 Intron NP_219482.1

View Full Product Details