Product Details

SNP ID
rs17088655
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:67820939 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGATTTGTTTGTACAGTTCGCCAA[G/T]AGTTGGTTGCATTTTCAACTCCCTT
Phenotype
MIM: 605369
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TMPRSS11D PubMed Links

Gene Details

Gene
TMPRSS11D
Gene Name
transmembrane protease, serine 11D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004262.2 2720 UTR 3 NP_004253.1
XM_005265710.4 2720 UTR 3 XP_005265767.1
XM_017008851.1 2720 UTR 3 XP_016864340.1

View Full Product Details