Product Details

SNP ID
rs17099573
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:100661797 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTTCTTTTCCTGACATCCAATGA[C/T]TCCCCACCTAATAGTGCATGGAGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00523 PubMed Links

Gene Details

Gene
LINC00523
Gene Name
long intergenic non-protein coding RNA 523
There are no transcripts associated with this gene.

View Full Product Details