Product Details

SNP ID
rs16835509
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:33480898 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATCCAGAACCTTCCTCTAAACATG[A/G]CTTTTTCTCTACTTGTTATAGTTAG
Phenotype
MIM: 611315
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZSCAN20 PubMed Links

Gene Details

Gene
ZSCAN20
Gene Name
zinc finger and SCAN domain containing 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145238.3 587 Intron NP_660281.2
XM_005271171.3 587 Intron XP_005271228.1
XM_006710874.3 587 Intron XP_006710937.1
XM_006710876.3 587 Intron XP_006710939.1
XM_017002237.1 587 Intron XP_016857726.1
XM_017002238.1 587 Intron XP_016857727.1
XM_017002239.1 587 Intron XP_016857728.1
XM_017002240.1 587 Intron XP_016857729.1
XM_017002241.1 587 UTR 5 XP_016857730.1

View Full Product Details