Product Details

SNP ID
rs17865426
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:90214671 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGGGAGTGACTACAAATCTAGCCT[C/T]GGTTTAAAGAGAAACCAGAGCAGGC
Phenotype
MIM: 605094
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STEAP2 PubMed Links

Gene Details

Gene
STEAP2
Gene Name
STEAP2 metalloreductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040665.1 2203 Intron NP_001035755.1
NM_001040666.1 2203 Intron NP_001035756.1
NM_001244944.1 2203 Intron NP_001231873.1
NM_001244945.1 2203 Intron NP_001231874.1
NM_001244946.1 2203 Intron NP_001231875.1
NM_152999.3 2203 Intron NP_694544.2
XM_006715921.3 2203 Intron XP_006715984.1
XM_017011953.1 2203 Intron XP_016867442.1
XM_017011954.1 2203 Intron XP_016867443.1
XM_017011955.1 2203 Intron XP_016867444.1
XM_017011956.1 2203 Intron XP_016867445.1
XM_017011957.1 2203 Intron XP_016867446.1
XM_017011958.1 2203 UTR 5 XP_016867447.1
Gene
STEAP2-AS1
Gene Name
STEAP2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details