Product Details

SNP ID
rs13447406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:5528088 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCAGGGTACATGGTGGTGCCGCC[A/G]GACAGCACTGTGTTGGCGTACAGGT
Phenotype
MIM: 102630
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACTB PubMed Links

Gene Details

Gene
ACTB
Gene Name
actin beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101.3 984 Silent Mutation NP_001092.1
Gene
LOC221946
Gene Name
uncharacterized LOC221946
There are no transcripts associated with this gene.

View Full Product Details