Product Details

SNP ID
rs17671851
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:35985587 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGTTGTGATTATTCTGACAGACTC[C/T]TCTTTAGACAGGCTTCTCTGAGCCC
Phenotype
MIM: 610347
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C18orf21 PubMed Links

Gene Details

Gene
C18orf21
Gene Name
chromosome 18 open reading frame 21
There are no transcripts associated with this gene.

Gene
RPRD1A
Gene Name
regulation of nuclear pre-mRNA domain containing 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303411.1 Intron NP_001290340.1
NM_001303412.1 Intron NP_001290341.1
NM_001303413.1 Intron NP_001290342.1
NM_018170.4 Intron NP_060640.2
XM_011526048.1 Intron XP_011524350.1
XM_011526049.1 Intron XP_011524351.1
XM_011526050.1 Intron XP_011524352.1
XM_017025808.1 Intron XP_016881297.1

View Full Product Details