Product Details
- SNP ID
-
rs17277546
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:99891948 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAAAAAGATTTCCCAAAAGCGTCTT[A/G]GAAAATTTCCTGTCCCATCTCCATT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TRIM4
PubMed Links
Gene Details
- Gene
- TRIM4
- Gene Name
- tripartite motif containing 4
View Full Product Details