Product Details

SNP ID
rs11541078
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:179796366 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCTGCGCGCCGCGCTCCTCGGAC[A/G]GCTCCGGACGCTGCTGCCGTGGGCC
Phenotype
MIM: 246530 MIM: 604561
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LTC4S PubMed Links

Gene Details

Gene
LTC4S
Gene Name
leukotriene C4 synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145867.1 521 Missense Mutation CAG,CGG Q142R NP_665874.1
Gene
MGAT4B
Gene Name
mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme B
There are no transcripts associated with this gene.

Gene
MIR1229
Gene Name
microRNA 1229
There are no transcripts associated with this gene.

View Full Product Details