Product Details

SNP ID
rs16912219
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5243049 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGCCACCTCTGCTTTCAAAAGTA[C/T]CCATAGTGCAAATATCTAAGGGTAA
Phenotype
MIM: 616308 MIM: 142000 MIM: 142200
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BGLT3 PubMed Links

Gene Details

Gene
BGLT3
Gene Name
beta globin locus transcript 3 (non-protein coding)
There are no transcripts associated with this gene.

Gene
HBBP1
Gene Name
hemoglobin subunit beta pseudogene 1
There are no transcripts associated with this gene.

Gene
HBD
Gene Name
hemoglobin subunit delta
There are no transcripts associated with this gene.

Gene
HBG1
Gene Name
hemoglobin subunit gamma 1
There are no transcripts associated with this gene.

View Full Product Details