Product Details

SNP ID
rs16968041
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:76259814 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGCAGGCAATGTCCACCATGAAC[C/T]GGATCAGGGTAAAGGGGTTCTCCCC
Phenotype
MIM: 608053
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ETFA PubMed Links

Gene Details

Gene
ETFA
Gene Name
electron transfer flavoprotein alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000126.3 Intron NP_000117.1
NM_001127716.1 Intron NP_001121188.1
Gene
TYRO3P
Gene Name
TYRO3P protein tyrosine kinase pseudogene
There are no transcripts associated with this gene.

View Full Product Details