Product Details

SNP ID
rs16973190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:71474598 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAATCATGGGGTTGAGGAGCTGCCA[C/T]GACTATAACTAAAGACTTTCCCATA
Phenotype
MIM: 194525
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF19 PubMed Links

Gene Details

Gene
ZNF19
Gene Name
zinc finger protein 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006961.3 2218 UTR 3 NP_008892.2

View Full Product Details