Product Details

SNP ID
rs16990299
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45514028 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATCCTCCTACCTGCAAGGCTGAC[A/G]CTAAGGGTGCAGGAGAATGGGGAAG
Phenotype
MIM: 613941
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPINT3 PubMed Links

Gene Details

Gene
SPINT3
Gene Name
serine peptidase inhibitor, Kunitz type 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006652.1 Intron NP_006643.1

View Full Product Details