Product Details

SNP ID
rs17790792
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6905602 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTTCAGGAGGAGGACTGAGCTTA[C/T]CTGACTCCAGAGCTTTCAGGAGGGA
Phenotype
MIM: 131360
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ENO2 PubMed Links

Gene Details

Gene
ENO2
Gene Name
enolase 2
There are no transcripts associated with this gene.

Gene
LRRC23
Gene Name
leucine rich repeat containing 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135217.1 189 UTR 5 NP_001128689.1
NM_006992.3 189 UTR 5 NP_008923.1
NM_201650.2 189 UTR 5 NP_964013.1

View Full Product Details