Product Details

SNP ID
rs16853105
Assay Type
Functionally tested
NCBI dbSNP Submissions
50
Location
Chr.1:204219389 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAAGAACATGCCTCTCCCAAAGGC[C/T]GGAGAGCAAGTGTGCCACTTGTGCC
Phenotype
MIM: 607771
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GOLT1A PubMed Links

Gene Details

Gene
GOLT1A
Gene Name
golgi transport 1A
There are no transcripts associated with this gene.

Gene
PLEKHA6
Gene Name
pleckstrin homology domain containing A6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014935.4 7118 UTR 3 NP_055750.2
XM_005244966.4 7118 UTR 3 XP_005245023.2
XM_005244967.4 7118 UTR 3 XP_005245024.2
XM_005244968.4 7118 UTR 3 XP_005245025.2
XM_006711212.3 7118 UTR 3 XP_006711275.1
XM_006711213.3 7118 UTR 3 XP_006711276.2
XM_006711214.3 7118 UTR 3 XP_006711277.2
XM_006711215.3 7118 UTR 3 XP_006711278.2
XM_006711217.3 7118 UTR 3 XP_006711280.2
XM_006711219.3 7118 UTR 3 XP_006711282.2
XM_006711221.2 7118 UTR 3 XP_006711284.1
XM_006711222.3 7118 UTR 3 XP_006711285.1
XM_011509295.2 7118 UTR 3 XP_011507597.1
XM_011509296.2 7118 UTR 3 XP_011507598.1
XM_011509297.2 7118 UTR 3 XP_011507599.1
XM_011509298.2 7118 UTR 3 XP_011507600.1
XM_011509299.2 7118 UTR 3 XP_011507601.2
XM_011509301.2 7118 UTR 3 XP_011507603.1
XM_011509304.2 7118 UTR 3 XP_011507606.1
XM_017000687.1 7118 UTR 3 XP_016856176.1
XM_017000688.1 7118 UTR 3 XP_016856177.1
XM_017000689.1 7118 UTR 3 XP_016856178.1
XM_017000690.1 7118 UTR 3 XP_016856179.1
XM_017000691.1 7118 UTR 3 XP_016856180.1
XM_017000692.1 7118 UTR 3 XP_016856181.1

View Full Product Details