Product Details

SNP ID
rs17691453
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:39251745 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTATCTACTTACTCTTTGTGCCT[C/T]CTCCAGTTAGTACCACACTAGAGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C15orf54 PubMed Links

Gene Details

Gene
C15orf54
Gene Name
chromosome 15 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302797.1 Intron NP_001289726.1
NM_207445.3 Intron NP_997328.1

View Full Product Details