Product Details

SNP ID
rs17015803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:119433855 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGCCCTGCATCACTGAAAGCTGG[C/T]GGAGATGCCGGGTGGTTAGGAATGA
Phenotype
MIM: 182098
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCTR PubMed Links

Gene Details

Gene
SCTR
Gene Name
secretin receptor
There are no transcripts associated with this gene.

Gene
TMEM37
Gene Name
transmembrane protein 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_183240.2 Intron NP_899063.2
XM_005263598.4 Intron XP_005263655.1
XM_006712300.3 Intron XP_006712363.1
XM_011510659.2 Intron XP_011508961.1

View Full Product Details