Product Details

SNP ID
rs6581079
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55683135 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGTTTCCCTGGGATGTTCACAGC[A/G]ATGTCCAGGAAGCAGCTTGTGCATG
Phenotype
MIM: 600536
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITGA7 PubMed Links

Gene Details

Gene
ITGA7
Gene Name
integrin subunit alpha 7
There are no transcripts associated with this gene.

Gene
METTL7B
Gene Name
methyltransferase like 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152637.2 Intron NP_689850.2

View Full Product Details