Product Details

SNP ID
rs17059546
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:173531911 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATATATTTCAGGTTTGCCGCGGC[C/T]GGGCGGTAGGAGACAGGGACTTAAA
Phenotype
MIM: 602407
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HAND2 PubMed Links

Gene Details

Gene
HAND2
Gene Name
heart and neural crest derivatives expressed 2
There are no transcripts associated with this gene.

Gene
HAND2-AS1
Gene Name
HAND2 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details