Product Details

SNP ID
rs17080034
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:169735526 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCAAAACTAATTAAATACAGAAGG[A/C]TCTAATGCCTAGAATAGTAGACTTG
Phenotype
MIM: 616614
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HPF1 PubMed Links

Gene Details

Gene
HPF1
Gene Name
histone PARylation factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017867.2 Intron NP_060337.2
XM_017008332.1 Intron XP_016863821.1

View Full Product Details