Product Details

SNP ID
rs17085814
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:51850402 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTTTTCTCCCTGGATGTGATGATG[A/T]CTTTTAATTGTTATCTGGGTCAGGT
Phenotype
MIM: 612977
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DCUN1D4 PubMed Links

Gene Details

Gene
DCUN1D4
Gene Name
defective in cullin neddylation 1 domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040402.2 Intron NP_001035492.1
NM_001287755.1 Intron NP_001274684.1
NM_001287757.1 Intron NP_001274686.1
NM_015115.3 Intron NP_055930.2
XM_005265731.3 Intron XP_005265788.1
XM_011534379.2 Intron XP_011532681.1
XM_011534380.1 Intron XP_011532682.1
XM_017007912.1 Intron XP_016863401.1
XM_017007913.1 Intron XP_016863402.1

View Full Product Details