Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001286790.1 | 5208 | UTR 3 | NP_001273719.1 | ||
NM_144571.2 | 5208 | UTR 3 | NP_653172.2 | ||
XM_011531806.2 | 5208 | UTR 3 | XP_011530108.1 | ||
XM_011531807.2 | 5208 | UTR 3 | XP_011530109.1 | ||
XM_011531808.2 | 5208 | UTR 3 | XP_011530110.1 | ||
XM_011531809.1 | 5208 | UTR 3 | XP_011530111.1 | ||
XM_011531810.1 | 5208 | UTR 3 | XP_011530112.1 | ||
XM_017007956.1 | 5208 | UTR 3 | XP_016863445.1 | ||
XM_017007957.1 | 5208 | UTR 3 | XP_016863446.1 |