Product Details

SNP ID
rs17107179
Assay Type
Functionally tested
NCBI dbSNP Submissions
38
Location
Chr.1:52354176 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCAATAGAGACCAGGGCCCGCATA[A/C]GAACAACTTCCCCAGGCAGGCAGTA
Phenotype
MIM: 603755
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CC2D1B PubMed Links

Gene Details

Gene
CC2D1B
Gene Name
coiled-coil and C2 domain containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032449.2 Intron NP_115825.1
XM_005270590.2 Intron XP_005270647.1
XM_005270591.2 Intron XP_005270648.1
XM_005270593.2 Intron XP_005270650.1
XM_006710423.2 Intron XP_006710486.1
XM_017000559.1 Intron XP_016856048.1
Gene
ZFYVE9
Gene Name
zinc finger FYVE-type containing 9
There are no transcripts associated with this gene.

View Full Product Details