Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001170794.1 | 9074 | UTR 3 | NP_001164265.1 | ||
NM_021813.3 | 9074 | UTR 3 | NP_068585.1 | ||
XM_005248758.4 | 9074 | UTR 3 | XP_005248815.1 | ||
XM_005248759.4 | 9074 | UTR 3 | XP_005248816.1 | ||
XM_011536039.2 | 9074 | Intron | XP_011534341.1 | ||
XM_011536040.2 | 9074 | Intron | XP_011534342.1 | ||
XM_017011156.1 | 9074 | UTR 3 | XP_016866645.1 | ||
XM_017011157.1 | 9074 | UTR 3 | XP_016866646.1 | ||
XM_017011158.1 | 9074 | UTR 3 | XP_016866647.1 | ||
XM_017011159.1 | 9074 | UTR 3 | XP_016866648.1 | ||
XM_017011160.1 | 9074 | UTR 3 | XP_016866649.1 | ||
XM_017011161.1 | 9074 | UTR 3 | XP_016866650.1 | ||
XM_017011162.1 | 9074 | UTR 3 | XP_016866651.1 | ||
XM_017011163.1 | 9074 | UTR 3 | XP_016866652.1 | ||
XM_017011164.1 | 9074 | UTR 3 | XP_016866653.1 | ||
XM_017011165.1 | 9074 | Intron | XP_016866654.1 | ||
XM_017011166.1 | 9074 | UTR 3 | XP_016866655.1 | ||
XM_017011167.1 | 9074 | Intron | XP_016866656.1 |