Product Details

SNP ID
rs17767151
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:46590852 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTCTAATATAGTGATGAAGAGTT[A/G]AAACAGGGAAGGGCAAACCTATCAG
Phenotype
MIM: 600153 MIM: 605857
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIGF PubMed Links

Gene Details

Gene
PIGF
Gene Name
phosphatidylinositol glycan anchor biosynthesis class F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002643.3 Intron NP_002634.1
NM_173074.2 Intron NP_775097.1
XM_005264369.2 Intron XP_005264426.1
XM_011532908.2 Intron XP_011531210.1
Gene
RHOQ
Gene Name
ras homolog family member Q
There are no transcripts associated with this gene.

View Full Product Details