Product Details

SNP ID
rs16947586
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:117453716 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATCCTGGTACAAGATCAAGTCTCA[A/G]AGAGAGCGAGAAATGAGAGGCTTGG
Phenotype
MIM: 610737
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KSR2 PubMed Links

Gene Details

Gene
KSR2
Gene Name
kinase suppressor of ras 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173598.4 16304 UTR 3 NP_775869.3
XM_011538224.2 16304 UTR 3 XP_011536526.1
XM_011538225.2 16304 UTR 3 XP_011536527.1
XM_011538226.2 16304 Intron XP_011536528.1
XM_011538228.2 16304 UTR 3 XP_011536530.1
XM_011538229.2 16304 Intron XP_011536531.1
XM_017019208.1 16304 Intron XP_016874697.1
XM_017019209.1 16304 Intron XP_016874698.1
XM_017019210.1 16304 UTR 3 XP_016874699.1

View Full Product Details