Product Details

SNP ID
rs17106187
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:50444957 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTGAACTCTCTGCTGTTCCCAGAA[A/G]CACTCAAATAGGGGCTTACTTGCAG
Phenotype
MIM: 604460
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAF1 PubMed Links

Gene Details

Gene
FAF1
Gene Name
Fas associated factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007051.2 Intron NP_008982.1
XM_017000136.1 Intron XP_016855625.1

View Full Product Details