Product Details
- SNP ID
-
rs17106184
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
29
- Location
-
Chr.1:50444313 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAAGAGCCTAAAAGCTATGTTCTCC[A/G]AGAGACAGCTAAAGAATGGGAGATA
- Phenotype
-
MIM: 604460
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FAF1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79278986] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAF1
- Gene Name
- Fas associated factor 1
View Full Product Details