Product Details

SNP ID
rs16838246
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:206451816 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACACAAGGGGGTCTGGATTTATTCT[A/G]TATCAAACTGTAAGTCCTAGGAAGT
Phenotype
MIM: 603710
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAM23 PubMed Links

Gene Details

Gene
ADAM23
Gene Name
ADAM metallopeptidase domain 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003812.3 Intron NP_003803.1
XM_005246932.3 Intron XP_005246989.1
XM_011512086.2 Intron XP_011510388.1

View Full Product Details