Product Details

SNP ID
rs16921315
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:95070041 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATCTTAAACGTGACCAAAATATGA[C/T]GTTAAAATCAGCAACTCTTTATACG
Phenotype
MIM: 603269
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SRSF8 PubMed Links

Gene Details

Gene
SRSF8
Gene Name
serine and arginine rich splicing factor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032102.3 3166 UTR 3 NP_115285.1
XM_017017142.1 3166 UTR 3 XP_016872631.1

View Full Product Details