Product Details

SNP ID
rs16845558
Assay Type
Functionally tested
NCBI dbSNP Submissions
32
Location
Chr.1:173187355 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGCAAAGGGTGGCAGTGGTGCCC[C/T]ATGCCTTAGCCCTGCACACTACCCA
Phenotype
MIM: 603594
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TNFSF4 PubMed Links

Gene Details

Gene
TNFSF4
Gene Name
tumor necrosis factor superfamily member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297562.1 Intron NP_001284491.1
NM_003326.4 Intron NP_003317.1
XM_011509964.2 Intron XP_011508266.2
XM_017002228.1 Intron XP_016857717.1
XM_017002229.1 Intron XP_016857718.1
XM_017002230.1 Intron XP_016857719.1

View Full Product Details