Product Details

SNP ID
rs35625644
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:129150650 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTCTATAAAATGGAGATCCTAA[C/T]GCCCTCTTGGGTTTGTGGTCAGAAT
Phenotype
MIM: 610120
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TSPAN33 PubMed Links

Gene Details

Gene
TSPAN33
Gene Name
tetraspanin 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178562.4 Intron NP_848657.1
XM_006715960.3 Intron XP_006716023.1

View Full Product Details