Product Details

SNP ID
rs16930922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:30014778 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAAAACGTAAACTTCCATTAACAA[C/T]GGCCCTTCACCAACAGTAAAATGGG
Phenotype
MIM: 614398
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA1462 PubMed Links

Gene Details

Gene
KIAA1462
Gene Name
KIAA1462
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020848.2 7389 UTR 3 NP_065899.1
XM_011519608.1 7389 UTR 3 XP_011517910.1
XM_011519609.2 7389 UTR 3 XP_011517911.1

View Full Product Details