Product Details

SNP ID
rs17678858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:13891971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACAAGCTCTGAAAAGGCTGCATG[A/G]TAGACAAAGTGGCTCATTTTAGAAA
Phenotype
MIM: 600541
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ETV1 PubMed Links

Gene Details

Gene
ETV1
Gene Name
ETS variant 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163147.1 5547 UTR 3 NP_001156619.1
NM_001163148.1 5547 UTR 3 NP_001156620.1
NM_001163149.1 5547 UTR 3 NP_001156621.1
NM_001163150.1 5547 UTR 3 NP_001156622.1
NM_001163151.1 5547 UTR 3 NP_001156623.1
NM_001163152.1 5547 UTR 3 NP_001156624.1
NM_004956.4 5547 UTR 3 NP_004947.2
XM_005249635.4 5547 Intron XP_005249692.1
XM_005249636.4 5547 Intron XP_005249693.1
XM_011515167.2 5547 Intron XP_011513469.1
XM_011515168.2 5547 Intron XP_011513470.1
XM_011515170.2 5547 Intron XP_011513472.1

View Full Product Details