Product Details

SNP ID
rs17153766
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59456991 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGATCAACCACACTAATGTGAAG[C/G]AGTTTTTCTTCCTGGAACTTACACG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR4D6 PubMed Links

Gene Details

Gene
OR4D6
Gene Name
olfactory receptor family 4 subfamily D member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004708.1 31 Missense Mutation CAG,GAG Q11E NP_001004708.1

View Full Product Details