Product Details
- SNP ID
-
rs17152533
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:79456803 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGTCCTAATAGTCAATACCCTTTGA[C/T]TGGATCTTAACTGATAAAAGTGGAA
- Phenotype
-
MIM: 606382
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MAGI2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79590625] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MAGI2
- Gene Name
- membrane associated guanylate kinase, WW and PDZ domain containing 2
There are no transcripts associated with this gene.
- Gene
- MAGI2-AS3
- Gene Name
- MAGI2 antisense RNA 3
There are no transcripts associated with this gene.
View Full Product Details