Product Details

SNP ID
rs16910453
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:14976457 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGTTATAGAATATGAGGTTTCAGG[A/T]ATATGCAGTGTGGCCAGTAAAGAGA
Phenotype
MIM: 601190
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PDE6H PubMed Links

Gene Details

Gene
PDE6H
Gene Name
phosphodiesterase 6H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006205.2 Intron NP_006196.1
XM_017019431.1 Intron XP_016874920.1

View Full Product Details