Product Details

SNP ID
rs17467356
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12239088 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATAAACCCATTCTCCAGAAGACAA[C/T]CCAAACAAGTCTTTCTCTAAACCTG
Phenotype
MIM: 194542
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928574 PubMed Links

Gene Details

Gene
LOC101928574
Gene Name
uncharacterized LOC101928574
There are no transcripts associated with this gene.

Gene
LOC107984139
Gene Name
zinc finger protein 791-like
There are no transcripts associated with this gene.

Gene
ZNF44
Gene Name
zinc finger protein 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164276.1 Intron NP_001157748.1
NM_016264.3 Intron NP_057348.3
XM_005259934.3 Intron XP_005259991.1
XM_011528062.2 Intron XP_011526364.1
XM_011528064.1 Intron XP_011526366.1
XM_011528066.1 Intron XP_011526368.1
XM_017026870.1 Intron XP_016882359.1
XM_017026871.1 Intron XP_016882360.1

View Full Product Details