Product Details

SNP ID
rs16986778
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:112084079 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCCACTGTACTTCCTGTTGCTCTT[C/T]AACCCCCTCTGCACGCCCTGGGAGC
Phenotype
MIM: 300334
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRPC5 PubMed Links

Gene Details

Gene
TRPC5
Gene Name
transient receptor potential cation channel subfamily C member 5
There are no transcripts associated with this gene.

Gene
ZCCHC16
Gene Name
zinc finger CCHC-type containing 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004308.2 Intron NP_001004308.2

View Full Product Details