Product Details

SNP ID
rs17325557
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:149607347 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGTTCTCCCCTCAACCCATCAGTA[C/T]TCAGCTTTGGGCCCTCAGCTTCTAT
Phenotype
MIM: 300031
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM185A PubMed Links

Gene Details

Gene
TMEM185A
Gene Name
transmembrane protein 185A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174092.2 Intron NP_001167563.1
NM_001282302.1 Intron NP_001269231.1
NM_032508.3 Intron NP_115897.1

View Full Product Details