Product Details

SNP ID
rs17145723
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73540493 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGACGGATTAGCACCTGGGAGTTC[C/T]GCCTGAAGGCTTAAAGTCACCATCA
Phenotype
MIM: 605846
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BCL7B PubMed Links

Gene Details

Gene
BCL7B
Gene Name
BCL tumor suppressor 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197244.1 Intron NP_001184173.1
NM_001301061.1 Intron NP_001287990.1
NM_001707.3 Intron NP_001698.2

View Full Product Details