Product Details

SNP ID
rs17851522
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:119300486 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCCTCCCTCGCTAAAGCCTCGC[C/T]GCTCCCGGAGACCATCTCGACCATC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C4orf3 PubMed Links

Gene Details

Gene
C4orf3
Gene Name
chromosome 4 open reading frame 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001701.3 449 Missense Mutation CAG,CGG Q17R NP_001001701.2
NM_001170330.1 449 Missense Mutation CAG,CGG Q150R NP_001163801.1
Gene
USP53
Gene Name
ubiquitin specific peptidase 53
There are no transcripts associated with this gene.

View Full Product Details