Product Details
- SNP ID
-
rs17057846
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:160475306 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTACAGAAAGGGCTAGCAAAGTTCA[A/G]TACTGGAGGATCACCTTACAGCTTC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR3142
PubMed Links
Gene Details
- Gene
- MIR3142
- Gene Name
- microRNA 3142
There are no transcripts associated with this gene.
- Gene
- MIR3142HG
- Gene Name
- MIR3142 host gene
There are no transcripts associated with this gene.
View Full Product Details