Product Details

SNP ID
rs17467420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:17922876 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATGACTCCAGTCTAGCTCGCATT[G/T]CGGCTCCCGCCCGGGCGAGTTCTCG
Phenotype
MIM: 600299
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PCM1 PubMed Links

Gene Details

Gene
PCM1
Gene Name
pericentriolar material 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001315507.1 114 UTR 5 NP_001302436.1
NM_001315508.1 114 UTR 5 NP_001302437.1
NM_006197.3 114 UTR 5 NP_006188.3
XM_006716336.3 114 UTR 5 XP_006716399.1
XM_006716340.3 114 UTR 5 XP_006716403.1
XM_006716349.3 114 UTR 5 XP_006716412.1
XM_011544528.2 114 UTR 5 XP_011542830.1
XM_011544529.2 114 UTR 5 XP_011542831.1
XM_011544530.2 114 UTR 5 XP_011542832.1
XM_011544532.2 114 UTR 5 XP_011542834.1
XM_011544533.2 114 UTR 5 XP_011542835.1
XM_011544534.2 114 UTR 5 XP_011542836.1
XM_011544535.2 114 UTR 5 XP_011542837.1
XM_011544540.2 114 UTR 5 XP_011542842.1
XM_017013472.1 114 UTR 5 XP_016868961.1
XM_017013473.1 114 UTR 5 XP_016868962.1
XM_017013474.1 114 UTR 5 XP_016868963.1
XM_017013475.1 114 Intron XP_016868964.1
XM_017013476.1 114 Intron XP_016868965.1
XM_017013477.1 114 UTR 5 XP_016868966.1
XM_017013478.1 114 UTR 5 XP_016868967.1
XM_017013479.1 114 UTR 5 XP_016868968.1
XM_017013480.1 114 UTR 5 XP_016868969.1
XM_017013481.1 114 UTR 5 XP_016868970.1
XM_017013482.1 114 UTR 5 XP_016868971.1
XM_017013483.1 114 UTR 5 XP_016868972.1
XM_017013484.1 114 Intron XP_016868973.1
XM_017013485.1 114 UTR 5 XP_016868974.1
XM_017013486.1 114 UTR 5 XP_016868975.1
XM_017013487.1 114 UTR 5 XP_016868976.1
XM_017013488.1 114 UTR 5 XP_016868977.1
XM_017013489.1 114 UTR 5 XP_016868978.1
XM_017013490.1 114 UTR 5 XP_016868979.1
XM_017013491.1 114 UTR 5 XP_016868980.1
XM_017013492.1 114 UTR 5 XP_016868981.1
XM_017013493.1 114 UTR 5 XP_016868982.1
XM_017013494.1 114 UTR 5 XP_016868983.1
XM_017013495.1 114 Intron XP_016868984.1
XM_017013496.1 114 Intron XP_016868985.1
XM_017013497.1 114 UTR 5 XP_016868986.1
XM_017013498.1 114 UTR 5 XP_016868987.1
XM_017013499.1 114 Intron XP_016868988.1
XM_017013500.1 114 UTR 5 XP_016868989.1
XM_017013501.1 114 UTR 5 XP_016868990.1
XM_017013502.1 114 Intron XP_016868991.1
XM_017013503.1 114 UTR 5 XP_016868992.1
XM_017013504.1 114 UTR 5 XP_016868993.1
XM_017013505.1 114 Intron XP_016868994.1
XM_017013506.1 114 UTR 5 XP_016868995.1
XM_017013507.1 114 UTR 5 XP_016868996.1
XM_017013508.1 114 Intron XP_016868997.1

View Full Product Details